Merck
Androgen receptor expression in meningiomas.
Journal of neurosurgery (1995-03-01)
R S Carroll, J Zhang, K Dashner, M Sar, E M Wilson, P M Black
RESUMO
The predominance of meningiomas in females, the accelerated growth of these tumors during the luteal phase of the menstrual cycle and during pregnancy, and the association between meningiomas and breast cancer have led to a number of studies examining the potential role of steroids in the growth of meningiomas. The possibility that androgens play a role in meningioma proliferation has been suggested by a small number of investigators. The aim of this study was to examine the expression of androgen receptor messenger ribonucleic acid (mRNA) and correlate it using immunochemistry with the nuclear localization of androgen receptor in a large number of meningiomas. Thirty-nine meningiomas were examined by Northern blot analysis for the presence of measurable amounts of androgen receptor mRNA and eight of these were analyzed by immunohistochemistry for receptor protein. Sixty-seven percent of the meningiomas expressed androgen receptor mRNA. There was a marked predominance of women among the patients whose tumors expressed androgen receptor; 69% were women and 31% were men. The immunohistochemical data correlated with Northern blot analysis of mRNA. The staining was predominantly nuclear, suggesting that the androgen receptor resides in a location that can activate gene expression.