Merck
  • Divergent regioselective synthesis of 2,5,6,7-tetrahydro-1H-1,4-diazepin-2-ones and 5H-1,4-benzodiazepines.
Divergent regioselective synthesis of 2,5,6,7-tetrahydro-1H-1,4-diazepin-2-ones and 5H-1,4-benzodiazepines.
The Journal of organic chemistry (2011-09-13)
Orazio A Attanasi, Lucia De Crescentini, Gianfranco Favi, Fabio Mantellini, Simona Nicolini
RESUMO
A novel and simple one-pot synthesis of 3-substituted 2,5,6,7-tetrahydro-1H-1,4-diazepin-2-ones from 1,2-diaza-1,3-dienes (DDs) and N-unsubstituted aliphatic 1,3-diamines is described. Here we also report a procedure to selectively obtain alkyl 5H-1,4-benzodiazepine-3-carboxylates from the DDs and 2-aminobenzylamine. Both processes occur by means of sequential 1,4-conjugated addition followed by regioselective 7-exo cyclization. The behavior of N-methyl- and N,N'-dimethyl-1,3-diaminopropanes toward the DDs furnished pyrazol-3-ones and bis-α-aminohydrazones, respectively.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Aminobenzylamine, 98%