Merck
  • Microspotting streptavidin and double-stranded DNA arrays on gold for high-throughput studies of protein-DNA interactions by surface plasmon resonance microscopy.
Microspotting streptavidin and double-stranded DNA arrays on gold for high-throughput studies of protein-DNA interactions by surface plasmon resonance microscopy.
Analytical chemistry (2004-02-14)
Jennifer S Shumaker-Parry, M Hadi Zareie, Ruedi Aebersold, Charles T Campbell
RESUMO
We present two strategies for microspotting 10 x 12 arrays of double-stranded DNAs (dsDNAs) onto a gold-coated glass slide for high-throughput studies of protein-DNA interactions by surface plasmon resonance (SPR) microscopy. Both methods use streptavidin (SA) as a linker layer between a biotin-containing mixed self-assembled monolayer (SAM) and biotinylated dsDNAs to produce arrays with high packing density. The primary mixed SAM is produced from biotin- and oligo(ethylene glycol)-terminated thiols bonded as thiolates onto the gold surface. In the first method, a robotic microspotter is used to deliver nanoliter droplets of dsDNA solution onto a uniform layer of this SA ( approximately 2 x 10(12) SA/cm(2)). SPR microscopy shows a density of (5-6) x 10(11) dsDNA/cm(2) (0.2-0.3 dsDNA/SA) in the array elements. The second method uses instead a microspotted array of this SA linker layer, onto which the microspots of dsDNA are added with spatial registry. SPR microscopy before addition of the dsDNA shows a SA coverage of 2 x 10(12) SA/cm(2) within the spots and a dsDNA density of 8.5 +/- 3.5 x 10(11) dsDNA/cm(2) (0.3-0.7 dsDNA/SA, depending on the length of dsDNA) after dsDNA spotting. We demonstrate the ability to simultaneously monitor protein binding with the SPR microscope in many 200-microm spots with 1-s time resolution and sensitivity to <1 pg of protein.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
SAM Formation reagent for biotin terminated monolayer on gold surface