Merck
  • Acid mine drainage in the Iberian Pyrite Belt: 1. Hydrochemical characteristics and pollutant load of the Tinto and Odiel rivers.
Acid mine drainage in the Iberian Pyrite Belt: 1. Hydrochemical characteristics and pollutant load of the Tinto and Odiel rivers.
Environmental science and pollution research international (2013-04-17)
Jose M Nieto, Aguasanta M Sarmiento, Carlos R Canovas, Manuel Olias, Carlos Ayora
RESUMO
Acid mine drainage in the Iberian Pyrite Belt is probably the worst case in the world of surface water pollution associated with mining of sulphide mineral deposits. The Iberian Pyrite Belt is located in SW Iberian Peninsula, and it has been mined during the last 4,500 years. The central and eastern part of the Iberian Pyrite Belt is drained by the Tinto and Odiel rivers, which receive most of the acidic leachates from the mining areas. As a result, the main channels of the Tinto and Odiel rivers are very rich in metals and highly acidic until reaching the Atlantic Ocean. A significant amount of the pollutant load transported by these two rivers is delivered during the rainy season, as is usual in rivers of Mediterranean climate regions. Therefore, in order to have an accurate estimation of the pollutant loads transported by the Tinto and Odiel rivers, a systematic sampling on a weekly basis and a high temporal resolution sampling of floods events were both performed. Results obtained show that metal fluxes are strongly dependent on the study period, highlighting the importance of inter-annual studies involving dry and wet years.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Iron disulfide, powder, −325 mesh, 99.8% trace metals basis