Merck
  • Synthesis and antiinflammatory activities of 3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones.
Synthesis and antiinflammatory activities of 3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones.
Journal of medicinal chemistry (1987-11-01)
H Ikuta, H Shirota, S Kobayashi, Y Yamagishi, K Yamada, I Yamatsu, K Katayama
RESUMO
A series of 3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-ones was synthesized and evaluated as candidate antiinflammatory/analgesic agents as well as dual inhibitors of prostaglandin and leukotriene synthesis. Some compounds that showed dual inhibitory activity were found to possess equipotent antiinflammatory activities to indomethacin, with reduced ulcerogenic effects. One of the compounds, N-methoxy-3-(3,5-di-tert-butyl-4-hydroxybenzylidene)pyrrolidin-2-o ne, was found to have a wider safety margin than indomethacin or piroxicam, and was selected for detailed evaluation as a candidate drug for clinical application.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2,4-Dibromobutyryl chloride, technical, ~90% (GC)