Merck
  • Two-generation effects of the chitin synthesis inhibitor, teflubenzuron, on the aquatic midge Chironomus riparius.
Two-generation effects of the chitin synthesis inhibitor, teflubenzuron, on the aquatic midge Chironomus riparius.
Ecotoxicology and environmental safety (2011-03-15)
Koffi T Tassou, Ralf Schulz
RESUMO
Teflubenzuron, a chitin synthesis inhibitor was used in a two-generation test with Chironomus riparius (Insecta) to assess effects over a full life cycle from the first-instar larvae in the parental (P) generation until emergence in the subsequent F1 generation. Sediment spiked with teflubenzuron ranging from 10 to 390.6 μg/kg sediment dry weight (dw) was used. EC₅₀-values for fecundity and fertility were 112.7 and 74.5 μg/kg dw, respectively. Significant adverse effects were observed compared to the solvent control for emergence rate (P < 0.01) and also for developmental rate. No observed effect concentrations values were lower for emergence rate in the F1 generation (62.5 μg/kg dw) than in the P generation (100 μg/kg dw), demonstrating that the F1 generation was more affected than the P generation. Thus, this two-generation test may help detecting population level effects as an amendment to the risk assessment for chronic exposures to endocrine disrupting compounds.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Teflubenzuron, PESTANAL®, analytical standard