Merck
A concise synthesis of the molecular framework of pleuromutilin.
Chemical communications (Cambridge, England) (2010-11-17)
Junjia Liu, Stephen D Lotesta, Erik J Sorensen
RESUMO
Two syntheses of the tricyclic carbon skeleton of pleuromutilin are reported. Diastereoselective 1,4-conjugate additions were used to elaborate bicyclic precursors at an early stage of each route, while ring-forming olefin metatheses were executed to complete the pleuromutilin carbon framework. The congeners prepared are appropriately functionalized to enable access to diverse pleuromutilin analogues.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Pleuromutilin, ≥95%