Merck
  • CRTH2-specific binding characteristics of [3H]ramatroban and its effects on PGD2-, 15-deoxy-Delta12, 14-PGJ2- and indomethacin-induced agonist responses.
CRTH2-specific binding characteristics of [3H]ramatroban and its effects on PGD2-, 15-deoxy-Delta12, 14-PGJ2- and indomethacin-induced agonist responses.
European journal of pharmacology (2005-11-01)
Hiromi Sugimoto, Michitaka Shichijo, Mitsuhiro Okano, Kevin B Bacon
RESUMO
We previously showed that ramatroban (Baynastrade mark), a thromboxane A(2) (TxA(2)) antagonist, had inhibited prostaglandin D(2) (PGD(2))-stimulated human eosinophil migration mediated through activation of chemoattractant receptor-homologous molecule expressed on Th2 cells (CRTH2). However, detailed pharmacological characterization of its inhibitory activity has not been described. In the present study, we showed that [(3)H]ramatroban bound to a single receptor site on CRTH2 transfectants with a similar K(d) value (7.2 nM) to a TxA(2) receptor (8.7 nM). We also demonstrated that ramatroban inhibited PGD(2)-, 15-deoxy-Delta(12, 14)-PGJ(2) (15d-PGJ(2))- and indomethacin-induced calcium responses on CRTH2 transfectants in a competitive manner with similar pA(2) values (8.5, 8.5, and 8.6, respectively). This is the first report showing the evidence for direct binding of ramatroban to CRTH2, revealing its competitive inhibitory effects and another interesting finding that PGD(2), indomethacin and 15d-PGJ(2) share the same binding site with ramatroban on CRTH2.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ramatroban, ≥98% (HPLC), solid