Merck
Long-lived near-infrared luminescent lanthanide complexes of imidodiphosphinate "shell" ligands.
Inorganic chemistry (2005-08-30)
Andrew P Bassett, Rik Van Deun, Peter Nockemann, Peter B Glover, Benson M Kariuki, Kristof Van Hecke, Luc Van Meervelt, Zoe Pikramenou
RESUMO
Near-infrared emitting complexes of Nd(III), Er(III), and Yb(III) based on hexacoordinate lanthanide ions with an aryl functionalized imidodiphosphinate ligand, tpip, have been synthesized and fully characterized. Three tpip ligands form a shell around the lanthanide with the ligand coordinating via the two oxygens leading to neutral complexes, Ln(tpip)3. In the X-ray crystal structures of Er(III) and Nd(III) complexes there is evidence of CH-pi interactions between the phenyl groups. Photophysical investigations of solution samples of the complexes demonstrate that all complexes exhibit relatively long luminescence lifetimes in nondeuteurated solvents. Luminescence studies of powder samples have also been recorded for examination of the properties of NIR complexes in the solid state for potential material applications. The results underline the effective shielding of the lanthanide by the twelve phenyl groups of the tpip ligands and the reduction of high-energy vibrations in close proximity to the lanthanide, both features important in the design of NIR emitting lanthanide complexes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Imidodiphosphate sodium salt, ≥97%