- Practical and high-yield syntheses of dihydromorphine from tetrahydrothebaine and efficient syntheses of (8S)-8-bromomorphide.
Practical and high-yield syntheses of dihydromorphine from tetrahydrothebaine and efficient syntheses of (8S)-8-bromomorphide.
The Journal of organic chemistry (2003-03-01)
Anna K Przybyl, Judith L Flippen-Anderson, Arthur E Jacobson, Kenner C Rice
PMID12608825
RESUMO
A practical method for the conversion of tetrahydrothebaine to dihydromorphine in 92% yield is described. The procedure should allow more efficient production of opium products and may be easily modified for large-scale synthesis. The conversion of codeine to (8S)-8-bromomorphide, a potentially valuable intermediate to 6-demethoxyoripavine and derivatives, is also described. The absolute configuration of (8S)-8-bromomorphide was determined by a single-crystal X-ray diffraction study of the hydrobromide salt.