Merck
  • Protein and thiol oxidation in cells exposed to peroxyl radicals is inhibited by the macrophage synthesised pterin 7,8-dihydroneopterin.
Protein and thiol oxidation in cells exposed to peroxyl radicals is inhibited by the macrophage synthesised pterin 7,8-dihydroneopterin.
Biochimica et biophysica acta (2002-08-17)
Sean Duggan, Christopher Rait, Aaron Platt, Steven Gieseg
RESUMO
Monocyte cells are exposed to a range of reactive oxygen species (ROS) when they are recruited to a site of inflammation. In this study, we have examined the damage caused to the monocyte-like cell line U937 by peroxyl radicals and characterised the protective effect of the macrophage synthesised compound 7,8-dihydroneopterin. Exposure of U937 cells to peroxyl radicals, generated by the thermolytic breakdown of 2,2'-azobis(amidinopropane) dihydrochloride (AAPH), resulted in the loss of cell viability as measured by thiazolyl blue (MTT) reduction, and lactate dehydrogenase (LDH) leakage. The major form of cellular damage observed was cellular thiol loss and the formation of reactive protein hydroperoxides. Peroxyl radical oxidation of the cells only caused a small increase in cellular lipid oxidation measured. Supplementation of the media with increasing concentrations of 7,8-dihydroneopterin significantly reduced the cellular thiol loss and inhibited the formation of the protein hydroperoxides. High performance liquid chromatography (HPLC) analysis showed 7,8-dihydroneopterin was oxidised by both peroxyl radicals and preformed protein hydroperoxides to predominately 7,8-dihydroxanthopterin. The possibility that 7,8-dihydroneopterin is a cellular antioxidant protecting macrophage proteins during inflammation is discussed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
7,8-Dihydroneopterin, ≥97.0% (HPLC)