Merck
Immunomodulation with DiNAC-- a new approach to the treatment of atherosclerosis?
Expert opinion on investigational drugs (2002-05-09)
Sheila A Doggrell
RESUMO
Not all antioxidants reduce atherosclerosis. This may be because atherosclerosis has an autoimmune, inflammatory pathogenesis. As probucol is both an antioxidant and an immunomodulatory drug, it may be the immunomodulatory effect that underlies its ability to reduce atherosclerosis. N,N-Diacetyl-L-cystine is not an antioxidant but is immunomodulatory. In the Watanabe-heritable hyperlipidaemic rabbit model of familial hypercholesterolaemia, N,N-diacetyl-L-cystine treatment does not lower lipid levels but it does reduce atherosclerosis. Immunomodulation may be a new approach to the treatment of atherosclerosis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Impureza C de acetilcisteína, European Pharmacopoeia (EP) Reference Standard