Merck
  • Evidence against the involvement of multiple radical generating sites in the expression of the vascular cell adhesion molecule-1.
Evidence against the involvement of multiple radical generating sites in the expression of the vascular cell adhesion molecule-1.
Free radical research (1998-06-30)
A Pietersma, N de Jong, L E de Wit, R G Kraak-Slee, J F Koster, W Sluiter
RESUMO
The present study was undertaken to investigate the hypothesis that multiple oxygen radical generating systems contribute to the tumor necrosis factor (TNF) alpha-stimulated transcriptional activation of the vascular cell adhesion molecule (VCAM)-1 in endothelial cells. Experimental evidence has implicated the cytochrome P450 monooxygenase and a phagocyte type NADPH-oxidase as a source of oxygen radicals in these cells. We show here that endothelial cells exhibit cytochrome P450 activity by measuring the O-dealkylation of the exogenous substrate 7-ethoxyresorufin, but components of the phagocyte-type NADPH oxidase could not be demonstrated in endothelial cells. In that latter respect it was surprising that the NADPH oxidase inhibitor apocynin completely prevented the accumulation of VCAM-1 mRNA. However, we found that apocynin also acts as an inhibitor of cytochrome P450 activity in endothelial cells. Therefore the inhibitory effect of apocynin on the induction of VCAM-1 may no longer be used to demonstrate a role for the NADPH oxidase in this process. Furthermore, different cytochrome P450 inhibitors Co2+, metyrapone, SKF525a decreased the endothelial VCAM-1 expression stimulated by TNFalpha. Also under hypoxic conditions the expression of VCAM-1 was reduced. On this basis we assume that the oxygen dependent step in the intracellular signalling cascade underlying the TNFalpha stimulated transcriptional activation of VCAM-1 resides in the activity of a cytochrome P450 dependent monooxygenase. The finding that the phospholipase A2 inhibitor bromophenacylbromide inhibited the expression of VCAM-1 may indicate that arachidonic acid serves as a substrate for the cytochrome P450 monooxygenase reaction, but further research is needed to elucidate the particular cytochrome P450 family member mediating the expression of VCAM-1.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Resorufin pentyl ether