Merck
Chiral high-performance liquid chromatographic determination of oxprenolol in plasma.
Journal of chromatography (1993-11-24)
M E Laethem, M T Rosseel, P Wijnant, F M Belpaire
RESUMO
A sensitive, stereospecific high-performance liquid chromatographic assay for oxprenolol enantiomers in rat plasma was developed, using a chiral derivatization agent. Racemic oxprenolol and the internal standard (racemic propranolol) are extracted with dichloromethane after alkalinization of the plasma. Quantitation of R(+)- and S(-)-oxprenolol is based on derivatization with the chiral agent S(-)-1-(1-naphthyl)-ethyl isocyanate, followed by chromatographic separation on a C18 reversed-phase column, with fluorometric detection (excitation at 226 nm, emission at 333 nm). The assay is reproducible as judged by a coefficient of variation of less than 17.5% for both enantiomers at all concentrations used. Preliminary experiments in the rat demonstrate that the method is sufficiently sensitive for pharmacokinetic studies in that species.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(R)-(−)-1-(1-Naphthyl)ethyl isocyanate, 98%