Merck
  • [Application of Pirkle-type chiral column to the resolution of enantiomeric drugs: resolution and identification of alpha-naphthyl isocyanate derivatives of alprenolol and some other beta-blockers].
[Application of Pirkle-type chiral column to the resolution of enantiomeric drugs: resolution and identification of alpha-naphthyl isocyanate derivatives of alprenolol and some other beta-blockers].
Yao xue xue bao = Acta pharmaceutica Sinica (1989-01-01)
Q Yang, Z P Sun, D K Ling
RESUMO
alpha-Naphthyl isocyanate (NI) as a derivatizing agent to separate several beta-blockers was described. This procedure is simple, rapid and gives good resolution. Experiments with NI derivatives of alprenolol showed that the isocyanate group reacted with the amino group on the analyte to form urea under mild conditions and with the hydroxyl group to form carbamate under relatively vigorous conditions. The results also indicated that the derivatized reactions is 1:1 between NT and analytes.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1-Naphthyl isocyanate, 98%