Merck
  • Clinically applicable antianginal agents suppress osteoblastic transformation of myogenic cells and heterotopic ossifications in mice.
Clinically applicable antianginal agents suppress osteoblastic transformation of myogenic cells and heterotopic ossifications in mice.
Journal of bone and mineral metabolism (2012-09-27)
Ryuichiro Yamamoto, Masaki Matsushita, Hiroshi Kitoh, Akio Masuda, Mikako Ito, Takenobu Katagiri, Tatsushi Kawai, Naoki Ishiguro, Kinji Ohno
RESUMO
Fibrodysplasia ossificans progressiva (FOP) is a rare autosomal dominant disorder characterized by progressive heterotopic ossification. FOP is caused by a gain-of-function mutation in ACVR1 encoding the bone morphogenetic protein type II receptor, ACVR1/ALK2. The mutant receptor causes upregulation of a transcriptional factor, Id1. No therapy is available to prevent the progressive heterotopic ossification in FOP. In an effort to search for clinically applicable drugs for FOP, we screened 1,040 FDA-approved drugs for suppression of the Id1 promoter activated by the mutant ACVR1/ALK2 in C2C12 cells. We found that that two antianginal agents, fendiline hydrochloride and perhexiline maleate, suppressed the Id1 promoter in a dose-dependent manner. The drugs also suppressed the expression of native Id1 mRNA and alkaline phosphatase in a dose-dependent manner. Perhexiline but not fendiline downregulated phosphorylation of Smad 1/5/8 driven by bone morphogenetic protein (BMP)-2. We implanted crude BMPs in muscles of ddY mice and fed them fendiline or perhexiline for 30 days. Mice taking perhexiline showed a 38.0 % reduction in the volume of heterotopic ossification compared to controls, whereas mice taking fendiline showed a slight reduction of heterotopic ossification. Fendiline, perhexiline, and their possible derivatives are potentially applicable to clinical practice to prevent devastating heterotopic ossification in FOP.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Perhexiline maleate salt, ≥98% (HPLC)