Merck
  • A general palladium-catalyzed carbonylative synthesis of 2-alkylbenzoxazinones from 2-bromoanilines and acid anhydrides.
A general palladium-catalyzed carbonylative synthesis of 2-alkylbenzoxazinones from 2-bromoanilines and acid anhydrides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-09-05)
Xiao-Feng Wu, Helfried Neumann, Matthias Beller
RESUMO
(C), its (O)K! An efficient palladium-catalyzed carbonylative synthesis of 2-alkylbenzoxazinones has been developed. By starting from 2-bromoanilines and acid anhydrides, the corresponding products were isolated in good yields.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-Bromoaniline, 98%