Merck
Regioselective diversification of a cardiac glycoside, lanatoside C, by organocatalysis.
The Journal of organic chemistry (2012-08-09)
Yoshihiro Ueda, Kenji Mishiro, Keisuke Yoshida, Takumi Furuta, Takeo Kawabata
RESUMO
Acylation of lanatoside C in the presence of organocatalyst 5 gave the C(4'''')-O-acylate in up to 90% regioselectivity (catalyst-controlled regioselectivity). Various functionalized acyl groups can be introduced at the C(4'''')-OH by a mixed anhydride method in the presence of 5 or the related organocatalyst. On the other hand, DMAP-catalyzed acylation of lanatoside C gave the C(3'''')-O-acylate in up to 97% regioselectivity (substrate-controlled regioselectivity). Thus, diverse regioselective introduction of acyl groups among eight free hydroxy groups of lanatoside C was achieved.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lanatoside C, ≥97% (TLC)