Merck
A chicory cytochrome P450 mono-oxygenase CYP71AV8 for the oxidation of (+)-valencene.
FEBS letters (2010-12-01)
Katarina Cankar, Adèle van Houwelingen, Dirk Bosch, Theo Sonke, Harro Bouwmeester, Jules Beekwilder
RESUMO
Chicory (Cichorium intybus L.), which is known to have a variety of terpene-hydroxylating activities, was screened for a P450 mono-oxygenase to convert (+)-valencene to (+)-nootkatone. A novel P450 cDNA was identified in a chicory root EST library. Co-expression of the enzyme with a valencene synthase in yeast, led to formation of trans-nootkatol, cis-nootkatol and (+)-nootkatone. The novel enzyme was also found to catalyse a three step conversion of germacrene A to germacra-1(10),4,11(13)-trien-12-oic acid, indicating its involvement in chicory sesquiterpene lactone biosynthesis. Likewise, amorpha-4,11-diene was converted to artemisinic acid. Surprisingly, the chicory P450 has a different regio-specificity on (+)-valencene compared to germacrene A and amorpha-4,11-diene.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(+)-Nootkatone, ≥99.0% (GC)