Merck
  • Preparation of molecularly imprinted solid phase extraction using bensulfuron-methyl imprinted polymer and clean-up for the sulfonylurea-herbicides in soybean.
Preparation of molecularly imprinted solid phase extraction using bensulfuron-methyl imprinted polymer and clean-up for the sulfonylurea-herbicides in soybean.
Analytica chimica acta (2008-04-15)
Kaijie Tang, Shangwei Chen, Xiaohong Gu, Haijun Wang, Jun Dai, Jian Tang
RESUMO
A pre-treatment methodology based on the molecularly imprinted solid phase extraction (MI-SPE) procedure was developed for the determination of bensulfuron-methyl (BSM), tribenuron-methyl (TBM), metsulfuron-methyl (MSM) and nicosulfuron (NS) in soybean samples. A molecular imprinted polymer (MIP) was prepared by precipitation polymerization using BSM as the template molecule, alpha-methacrylic acid (MAA) as the functional monomer, trimethylolpropane trimethacrylate (TRIM) as the cross-linker and dichloromethane as the porogen. The binding behaviors of the template BSM and its analogues on the MIP were evaluated by high performance liquid chromatography (HPLC). Then, solid phase extraction (SPE) with a BSM molecularly imprinted polymer (BSM-MIP) as adsorbent was investigated and the optimum loading, washing, and eluting conditions for MI-SPE of the selected BSM, MSM, TBM, and NS were established. The optimized MI-SPE procedure was used to extract the sulfonylureas and a high recovery was obtained in the soybean samples.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Bensulfuron-methyl, PESTANAL®, analytical standard