Merck
Reaction of isatin with thiocarbohydrazide: a correction.
Archiv der Pharmazie (1991-06-01)
M A Badawy, S A Abdel-Hady
RESUMO
Heterocycles containing the indole ring system include some novel pharmacologically active compounds. Isatin and its N-acetylisatin are extremely versatile intermediates in the construction of a variety of heterocyclic systems when reacted with thiosemicarbazide derivatives. Literature survey revealed various interesting reactions of thiocarbohydrazide with cyclic ketones, cyclic 1,2-diketones and isatin.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Thiocarbohydrazide, 98%