Merck
Suppression of humoral immunity following exposure to the perfluorinated insecticide sulfluramid.
Journal of toxicology and environmental health. Part A (2007-06-15)
Margie M Peden-Adams, Jackie G EuDaly, Sundeep Dabra, Amy EuDaly, Lauren Heesemann, Joshua Smythe, Deborah E Keil
RESUMO
Perfluorinated hydrocarbons have been manufactured for over 40 yr and have numerous applications in industry. This group of compounds has recently generated much interest, as some of these compounds such as perfluorooctane sulfonate (PFOS) and perfluoroctanic acid (PFOA) are persistent in the environment and detectable in blood samples of both wildlife and humans. Studies show that these perfluorinated compounds induce peroxisomal proliferation, induce hepatomegaly, alter steroidogenesis, and decrease body weight, accompanied by a wasting syndrome; however, effects on immune function have not been addressed at length. This study examined sulfluramid, a perfluorinated pesticide that is currently available in the marketplace and is a representative member of this class of chemicals. Adult female B6C3F1 mice were exposed via gavage to either an oil carrier control or sulfluramid for 14 d (1, 3, 10, or 30 mg/kg/d) or 28 d (0.3, 1, 3, or 10 mg/kg/d). Although responses were normal in natural killer cell activity and lymphocyte proliferation, dose-responsive suppression was noted in the plaque forming cell (PFC) response at exposure levels as low as 3 mg/kg/d in the 14-d exposure and 0.3 mg/kg/d for 28 d. Dose-responsive increases in liver mass were observed following treatment with 1, 3, 10, or 30 mg/kg/d for 14 d and 0.3, 1, 3, or 10 mg/kg/d for 28 d. A significant reduction in body weight was observed at the highest dose level in each study. Novel findings in this study indicate that sulfluramid suppresses immunoglobulin (Ig) M production. Additional immunotoxicity studies are required to understand potential mechanisms of suppression and determine potential health risks associated with exposure to perfluorinated hydrocarbons.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Sulfluramid, PESTANAL®, analytical standard