Merck
Military aviators, special operations forces, and causal malaria prophylaxis.
Military medicine (2004-01-15)
James A Chambers
RESUMO
U.S. military aviators are currently restricted to the use of chloroquine or doxycycline for malaria prophylaxis. Ground forces are allowed the additional option of taking mefloquine. These medications are begun before deployment, must be taken for 4 weeks after leaving the malarious area, and primaquine must be added to the regimen the last 2 of those 4 weeks. Compliance with this regimen is often poor, especially in populations who travel abroad frequently for short periods of time. Causal malaria prophylaxis offers potential benefits of decreased length of postdeployment regimens and obviates the need for a second medication for terminal prophylaxis. Potential obstacles include adverse drug reactions, cost, and rapid development of resistance to new medications by Plasmodium species, which should be weighed against the risks to health and mission success in each deployment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Tafenoquine succinate, ≥95% (HPLC)