Merck
  • Determination of isoxaflutole (balance) and its metabolites in water using solid phase extraction followed by high-performance liquid chromatography with ultraviolet or mass spectrometry.
Determination of isoxaflutole (balance) and its metabolites in water using solid phase extraction followed by high-performance liquid chromatography with ultraviolet or mass spectrometry.
Journal of agricultural and food chemistry (2002-10-03)
Chung-Ho Lin, Robert N Lerch, E Michael Thurman, Harold E Garrett, Milon F George
RESUMO
Balance (isoxaflutole, IXF) belongs to a new family of herbicides referred to as isoxazoles. IXF has a very short soil half-life (<24 h), degrading to a biologically active diketonitrile (DKN) metabolite that is more polar and considerably more stable. Further degradation of the DKN metabolite produces a nonbiologically active benzoic acid (BA) metabolite. Analytical methods using solid phase extraction followed by high-performance liquid chromatography-UV (HPLC-UV) or high-performance liquid chromatography-mass spectrometry (HPLC-MS) were developed for the analysis of IXF and its metabolites in distilled deionized water and ground water samples. To successfully detect and quantify the BA metabolite by HPLC-UV from ground water samples, a sequential elution scheme was necessary. Using HPLC-UV, the mean recoveries from sequential elution of the parent and its two metabolites from fortified ground water samples ranged from 68.6 to 101.4%. For HPLC-MS, solid phase extraction of ground water samples was performed using a polystyrene divinylbenzene polymer resin. The mean HPLC-MS recoveries of the three compounds from ground water samples spiked at 0.05-2 microg/L ranged from 100.9 to 110.3%. The limits of quantitation for HPLC-UV are approximately 150 ng/L for IXF, 100 ng/L for DKN, and 250 ng/L for BA. The limit of quantitation by HPLC-MS is 50 ng/L for each compound. The methods developed in this work can be applied to determine the transport and fate of Balance in the environment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Isoxaflutole, PESTANAL®, analytical standard