Merck
TALL-1 is a novel member of the TNF family that is down-regulated by mitogens.
Journal of leukocyte biology (1999-05-20)
H B Shu, W H Hu, H Johnson
RESUMO
Members of the tumor necrosis factor (TNF) family play important roles in modulation of immune responses. We describe the identification and cloning of a novel TNF family member that has been designated as TALL-1. TALL-1 is a 285-amino acid type II transmembrane protein. Its carboxy terminus shares approximately 35% sequence identity with the recently identified APRIL and approximately 20-25% with TNF, FasL, TRAIL, and lymphotoxin-alpha, suggesting that TALL-1 and APRIL belong to a subfamily of the TNF family of ligands. Northern blot analysis suggests that TALL-1 is expressed abundantly in peripheral blood leukocytes and weakly in spleen but is barely detectable in all other tissues examined. Reverse transcriptase-polymerase chain reaction analysis indicates that TALL-1 is specifically expressed in monocytes and macrophages but is undetectable in T and B lymphocytes. Furthermore, TALL-1 expression is dramatically down-regulated by phorbol myristate acetate/ionomycin.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
APRIL human, recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
APRIL from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture