Merck
The δ opioid receptor agonist SNC80 selectively activates heteromeric μ-δ opioid receptors.
ACS chemical neuroscience (2012-08-04)
Matthew D Metcalf, Ajay S Yekkirala, Michael D Powers, Kelley F Kitto, Carolyn A Fairbanks, George L Wilcox, Philip S Portoghese
RESUMO
Coexpressed and colocalized μ- and δ-opioid receptors have been established to exist as heteromers in cultured cells and in vivo. However the biological significance of opioid receptor heteromer activation is less clear. To explore this significance, the efficacy of selective activation of opioid receptors by SNC80 was assessed in vitro in cells singly and coexpressing opioid receptors using a chimeric G-protein-mediated calcium fluorescence assay, SNC80 produced a substantially more robust response in cells expressing μ-δ heteromers than in all other cell lines. Intrathecal SNC80 administration in μ- and δ-opioid receptor knockout mice produced diminished antinociceptive activity compared with wild type. The combined in vivo and in vitro results suggest that SNC80 selectively activates μ-δ heteromers to produce maximal antinociception. These data contrast with the current view that SNC80 selectively activates δ-opioid receptor homomers to produce antinociception. Thus, the data suggest that heteromeric μ-δ receptors should be considered as a target when SNC80 is employed as a pharmacological tool in vivo.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
SNC80, ≥98% (HPLC), solid