Merck
  • Purification and preparation of Rhodobacter sphaeroides reaction centers for photocurrent measurements and atomic force microscopy characterization.
Purification and preparation of Rhodobacter sphaeroides reaction centers for photocurrent measurements and atomic force microscopy characterization.
STAR protocols (2022-01-04)
Daniel Jun, Sylvester Zhang, Adrian Jan Grzędowski, Amita Mahey, J Thomas Beatty, Dan Bizzotto
RESUMO
The formation of defined surfaces consisting of photosynthetic reaction centers (RCs) in biohybrid solar cells is challenging. Here, we start with the production of engineered RCs for oriented binding. RCs are deposited onto gold electrodes, and 6-mercapto-1-hexanol (MCH) is used to displace multilayers and non-specifically adsorbed RCs. The resulting electrode surfaces are analyzed for photocurrent generation using an intensity-modulated light and lock-in amplifier. Atomic force microscopy (AFM) is used to characterize the surface and the formation of RC structural assemblies. For complete details on the use and execution of this profile, please refer to Jun et al. (2021).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N,N-Dimetildodecilamina-N-óxido, ~30% in H2O