Merck
  • Combined Effects of Nanoroughness and Ions Produced by Electrodeposition of Mesoporous Bioglass Nanoparticle for Bone Regeneration.
Combined Effects of Nanoroughness and Ions Produced by Electrodeposition of Mesoporous Bioglass Nanoparticle for Bone Regeneration.
ACS applied bio materials (2019-11-18)
Kapil D Patel, Jennifer O Buitrago, S Prakash Parthiban, Jung-Hwan Lee, Rajendra K Singh, Jonathan C Knowles, Hae-Won Kim
RESUMO
Providing appropriate biophysical and biochemical cues to the interface is a facile strategy to enhance the osteogenic ability of metallic implants. Here we exploited this through the incorporation of mesoporous bioactive glass nanoparticles (MBGN) at a high content (1:1 by weight) to a biopolymer chitosan in the electrodeposition process of titanium. The MGBN/chitosan layer thickness, tunable by electrodeposition parameters, exhibited an accelerated ability of apatite mineral induction in a body simulating medium. Of note, the involvement of MBGN could generate nanoscale roughness in a unique range of 10-25 nm. Moreover, the layer showed a slowly releasing profile of ions (calcium and silicate) over weeks at therapeutically relevant doses. The ion-releasing nanotopological surface was demonstrated to alter the preosteoblasts responses in a way favorable for osteogenic differentiation. The combinatory cues of nanotopology (25 nm roughness) and ion release enabled highly accelerated cellular anchorage with somewhat limited spreading area at initial periods. The subsequent osteoblastic differentiation behaviors on the engineered surface, as examined up to 21 days, showed significantly enhanced alkaline phosphate activity and up-regulated expression of bone-associated genes (ALP, Col I, OPN, and OCN). These results indicate that the combinatory cues provided by nanotopology (25 nm roughness) and ions released from MBGN are highly effective in stimulating osteoblastic differentiation and suggest that the MBGN/chitosan may serve as a potential composition for bone implant coatings.