Merck
The GTPase Rab27b regulates the release, autophagic clearance, and toxicity of α-synuclein.
The Journal of biological chemistry (2020-05-01)
Rachel Underwood, Bing Wang, Christine Carico, Robert H Whitaker, William J Placzek, Talene A Yacoubian, Rachel Underwood, Bing Wang, Christine Carico, Robert H Whitaker, William J Placzek, Talene A Yacoubian
RESUMO
α-Synuclein (αsyn) is the primary component of proteinaceous aggregates termed Lewy bodies that pathologically define synucleinopathies including Parkinson's disease (PD) and dementia with Lewy bodies (DLB). αsyn is hypothesized to spread through the brain in a prion-like fashion by misfolded protein forming a template for aggregation of endogenous αsyn. The cell-to-cell release and uptake of αsyn are considered important processes for its prion-like spread. Rab27b is one of several GTPases essential to the endosomal-lysosomal pathway and is implicated in protein secretion and clearance, but its role in αsyn spread has yet to be characterized. In this study, we used a paracrine αsyn in vitro neuronal model to test the impact of Rab27b on αsyn release, clearance, and toxicity. shRNA-mediated knockdown (KD) of Rab27b increased αsyn-mediated paracrine toxicity. Rab27b reduced αsyn release primarily through nonexosomal pathways, but the αsyn released after Rab27b KD was of higher-molecular-weight species, as determined by size-exclusion chromatography. Rab27b KD increased intracellular levels of insoluble αsyn and led to an accumulation of endogenous light chain 3 (LC3)-positive puncta. Rab27b KD also decreased LC3 turnover after treatment with an autophagosome-lysosome fusion inhibitor, chloroquine, indicating that Rab27b KD induces a defect in autophagic flux. Rab27b protein levels were increased in brain lysates obtained from postmortem tissues of individuals with PD and DLB compared with healthy controls. These data indicate a role for Rab27b in the release, clearance, and toxicity of αsyn and, ultimately, in the pathogenesis of synucleinopathies.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-α-tubulina monoclonal, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Rab27B Antibody, serum, from rabbit