Merck
  • New novobiocin analogues as antiproliferative agents in breast cancer cells and potential inhibitors of heat shock protein 90.
New novobiocin analogues as antiproliferative agents in breast cancer cells and potential inhibitors of heat shock protein 90.
Journal of medicinal chemistry (2007-11-06)
Gaëlle Le Bras, Christine Radanyi, Jean-François Peyrat, Jean-Daniel Brion, Mouâd Alami, Véronique Marsaud, Barbara Stella, Jack-Michel Renoir
RESUMO
Selective hsp90 inhibitors simultaneously destabilize and deplete key signaling proteins involved in cell proliferation and survival, angiogenesis, and metastasis. Investigation of novobiocin analogues lacking the noviose moiety as novel inhibitors of hsp90 was carried out. A novel series of 3-aminocoumarin analogues has been produced and screened in cell proliferation, and the molecular signature of hsp90 inhibition was assessed by depletion of estrogen receptor, HER2, Raf-1, and cdk4 in human breast cancer cells. This structure-activity relationship study highlights the crucial role of the C-4 and/or C-7 positions of coumarin which appeared to be essential for degradation of hsp90 client proteins. Removal of the noviose moiety in novobiocin together with introduction of a tosyl substituent at C-4 or C-7 coumarins provides 6e and 6f as lead structures which compared favorably with novobiocin as demonstrated by enhanced rates of cell death. The processing and activation of caspases 7 and 8 and the subsequent cleavage of PARP by 6e suggest stimulation of the extrinsic apoptosis pathway.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
18β-Glycyrrhetinic acid, 97%