Merck
  • Secretome-derived isotope tags (SDIT) reveal adipocyte-derived apolipoprotein C-I as a predictive marker for cardiovascular disease.
Secretome-derived isotope tags (SDIT) reveal adipocyte-derived apolipoprotein C-I as a predictive marker for cardiovascular disease.
Journal of proteome research (2012-03-13)
Rong-Xia Li, Yu-Bo Ding, Shi-Lin Zhao, Yuan-Yuan Xiao, Qing-run Li, Fang-Ying Xia, Liang Sun, Xu Lin, Jia-Rui Wu, Kan Liao, Rong Zeng
RESUMO
We developed a quantitative strategy, named secretome-derived isotopic tag (SDIT), to concurrently identify and quantify the adipocyte-secreted plasma proteins from normal and high-fat-diet (HFD) induced obese mice, based on the application of isotope-labeled secreted proteins from cultured mouse adipocytes as internal standards. We detected 197 proteins with significant changes between normal and obese mice plasma. Importantly, a novel adipocyte-secreted plasma protein, apolipoprotein C-I (apoC-I), significantly increased in the obese mice plasma. The expression and secretion of adipocyte apoC-I was detected in differentiated 3T3-L1 and primary rat adipocytes. Our in vitro experiments proved that functional Golgi apparatus was required for apoC-I secretion. Additionally, obese mice had increased apoC-I production in adipose tissue. Population survey of 367 participants showed that the plasma level of apoC-I was significantly increased in obese individuals compared with healthy individuals. After multiple adjustments for age and sex, the odds ratios for risk factors of cardiovascular disease including high LDL cholesterol, hypercholesterolemia, and hypertriglyceridemia, respectively, were used to compare the highest with the lowest apoC-I quartile. Taken together, our studies provide a novel strategy to concurrently identify and quantify tissue-specific secreted proteins. This strategy can be used to identify the largest global characterization of adipocyte-derived plasma proteome and provides a potential disease-related biomarker for clinical diagnoses. By selectively analyzing adipocyte-secreted proteins in plasma from obese vs lean murine and/or human subjects, we discovered that apoC-I is an adipocyte-secreted plasma protein and a predictive marker for cardiovascular disease.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Apolipoprotein C-I from human plasma, ≥95% (SDS-PAGE), lyophilized powder