Merck
  • Site-directed mutagenesis of CC chemokine receptor 1 reveals the mechanism of action of UCB 35625, a small molecule chemokine receptor antagonist.
Site-directed mutagenesis of CC chemokine receptor 1 reveals the mechanism of action of UCB 35625, a small molecule chemokine receptor antagonist.
The Journal of biological chemistry (2004-11-19)
Filipa Lopes de Mendonça, Paula C A da Fonseca, Rhian M Phillips, José W Saldanha, Timothy J Williams, James E Pease
RESUMO
The chemokine receptor CCR1 and its principal ligand, CCL3/MIP-1alpha, have been implicated in the pathology of several inflammatory diseases including rheumatoid arthritis, multiple sclerosis, and asthma. As such, these molecules are the focus of much research with the ultimate aim of developing novel therapies. We have described previously a non-competitive small molecule antagonist of CCR1 (UCB 35625), which we hypothesized interacted with amino acids located within the receptor transmembrane (TM) helices (Sabroe, I., Peck, M. J., Jan Van Keulen, B., Jorritsma, A., Simmons, G., Clapham, P. R., Williams, T. J., and Pease, J. E. (2000) J. Biol. Chem. 275, 25985-25992). Here we describe an approach to identifying the mechanism by which the molecule antagonizes CCR1. Thirty-three point mutants of CCR1 were expressed transiently in L1.2 cells, and the cells were assessed for their capacity to migrate in response to CCL3 in the presence or absence of UCB 35625. Cells expressing the mutant constructs Y41A (TM helix 1, or TM1), Y113A (TM3), and E287A (TM7) were responsive to CCL3 but resistant to the antagonist, consistent with a role for the TM helices in CCR1 interactions with UCB 35625. Subsequent molecular modeling successfully docked the compound with CCR1 and suggests that the antagonist ligates TM1, 2, and 7 of CCR1 and severely impedes access to TM2 and TM3, a region thought to be perturbed by the chemokine amino terminus during the process of receptor activation. Insights into the mechanism of action of these compounds may facilitate the development of more potent antagonists that show promise as future therapeutic agents in the treatment of inflammatory disease.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
UCB35625, >97% (HPLC)