Merck
Free fatty acids decrease circulating ghrelin concentrations in humans.
European journal of endocrinology (2006-04-29)
Lars C Gormsen, Jakob Gjedsted, Signe Gjedde, Esben Thyssen Vestergaard, Jens S Christiansen, Jens Otto Jørgensen, Søren Nielsen, Niels Møller
RESUMO
Concentrations of the orexigenic peptide ghrelin is affected by a number of hormones, which also affect circulating levels of free fatty acids (FFAs). The present study was therefore designed to determine the direct effect of FFAs on circulating ghrelin. Eight lean, healthy men were examined for 8 h on four occasions using variable infusion rates (0, 3, 6 and 12 microl/kg per min) of intralipid to create different plasma FFA concentrations. Constant levels of insulin and GH were obtained by administration of acipimox (250 mg) and somatostatin (300 microg/h). At the end of each study day a hyperinsulinaemic-euglycaemic clamp was performed. Four distinct levels of FFAs were obtained at the end of the lipid infusion period (FFA(LIPID): 0.03 +/- 0.00 vs: 0.49 +/- 0.04, 0.92 +/- 0.08 and 2.09 +/- 0.38 mmol/l; ANOVA P < 0.0001) and during hyperinsulinaemia (FFA(LIPID+INSULIN): 0.02 +/- 0.00 vs: 0.34 +/- 0.03, 0.68 +/- 0.09 and 1.78 +/- 0.32 mmol/l; ANOVA P < 0.0001). Whereas, somatostatin infusion alone reduced ghrelin concentration by approximately 67%, concomitant administration of increasing amounts of intralipid reduced circulating ghrelin by a further 14, 19 and 19% respectively (change in ghrelin: 0.52 +/- 0.05 vs: 0.62 +/- 0.06, 0.72 +/- 0.09 and 0.71 +/- 0.05 microg/l; ANOVA P = 0.04). No further reduction in ghrelin concentration was observed during hyperinsulinaemia. FFA exposure between 0 and 1 mmol/l significantly suppresses ghrelin levels independent of ambient GH and insulin levels.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acipimox, ≥99% (TLC)
SKU
Tamanho da embalagem
Disponibilidade
Preço
Quantidade