Merck
  • Synthesis of L-thiocitrulline, L-homothiocitrulline, and S-methyl-L-thiocitrulline: a new class of potent nitric oxide synthase inhibitors.
Synthesis of L-thiocitrulline, L-homothiocitrulline, and S-methyl-L-thiocitrulline: a new class of potent nitric oxide synthase inhibitors.
Journal of medicinal chemistry (1994-04-01)
K Narayanan, O W Griffith
RESUMO
Nitric oxide synthase catalyzes the NADPH- and O2-dependent conversion of L-arginine to L-citrulline and nitric oxide. L-Thiocitrulline, L-homothiocitrulline, and S-methyl-L-thiocitrulline, novel citrulline analogs, have been synthesized and are shown to be potent inhibitors of both the constitutive brain and the inducible smooth muscle isoforms of nitric oxide synthase. Although many N omega-monosubstituted arginine derivatives inhibit nitric oxide synthase, inhibitory citrulline derivatives have not previously been reported. S-Methyl-L-thiocitrulline is significantly more potent than N omega-methyl-L-arginine, the prototypic nitric oxide synthase inhibitor.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
S-Methyl-L-thiocitrulline acetate salt, ≥98% (TLC)