Merck
Redundant mitochondrial targeting signals in yeast adenylate kinase.
The Journal of biological chemistry (2002-06-05)
Roland Schricker, Michaela Angermayr, Gertrud Strobel, Sigrid Klinke, Dorothee Korber, Wolfhard Bandlow
RESUMO
Yeast adenylate kinase (Aky2p, Adk1p) occurs simultaneously in cytoplasm and mitochondrial intermembrane space. It has no cleavable mitochondrial targeting sequence, and the signal for mitochondrial import and submitochondrial sorting is largely unknown. The extreme N terminus of Aky2p is able to direct cytoplasmic passengers to mitochondria. However, an Aky2 mutant lacking this sequence is imported with about the same efficiency as the wild type. To identify possible import-relevant information in the interior, parts of Aky2p were exchanged by homologous in vitro recombination for the respective segments of the purely cytoplasmic isozyme, Ura6p. Import studies revealed an internal region of about 40 amino acids, which was sufficient to direct the chimera to mitochondria but not for correct submitochondrial sorting. The respective Ura6p hybrid was arrested in the mitochondrial membrane at a position where it was inaccessible to protease but was released by alkaline extraction, suggesting that it had entered an import channel and passed the initial steps of recognition and uptake. Site-specific mutations within the presumptive address-specifying segment identified the amphipathic helix 5. A Ura6 mutant protein in which helix 5 had been replaced with the respective sequence from Aky2p was imported, and this address sequence cooperates with the N terminus in the respective double mutant in a synergistic fashion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Myokinase, Yeast