MilliporeSigma
A KDM6A-KLF10 reinforcing feedback mechanism aggravates diabetic podocyte dysfunction.
EMBO molecular medicine (2019-04-06)
Chun-Liang Lin, Yung-Chien Hsu, Yu-Ting Huang, Ya-Hsueh Shih, Ching-Jen Wang, Wen-Chih Chiang, Pey-Jium Chang
RESUMO
Diabetic nephropathy is the leading cause of end-stage renal disease. Although dysfunction of podocytes, also termed glomerular visceral epithelial cells, is critically associated with diabetic nephropathy, the mechanism underlying podocyte dysfunction still remains obscure. Here, we identify that KDM6A, a histone lysine demethylase, reinforces diabetic podocyte dysfunction by creating a positive feedback loop through up-regulation of its downstream target KLF10. Overexpression of KLF10 in podocytes not only represses multiple podocyte-specific markers including nephrin, but also conversely increases KDM6A expression. We further show that KLF10 inhibits nephrin expression by directly binding to the gene promoter together with the recruitment of methyltransferase Dnmt1. Importantly, inactivation or knockout of either KDM6A or KLF10 in mice significantly suppresses diabetes-induced proteinuria and kidney injury. Consistent with the notion, we also show that levels of both KDM6A and KLF10 proteins or mRNAs are substantially elevated in kidney tissues or in urinary exosomes of human diabetic nephropathy patients as compared with control subjects. Our findings therefore suggest that targeting the KDM6A-KLF10 feedback loop may be beneficial to attenuate diabetes-induced kidney injury.