Merck
Hypophagic effects of insulin are mediated via NPY1/NPY2 receptors in broiler cockerels.
Canadian journal of physiology and pharmacology (2018-10-17)
Shiba Yousefvand, Farshid Hamidi, Morteza Zendehdel, Abbas Parham
RESUMO
Neuropeptide Y (NPY) plays a mediatory role in cerebral insulin function by maintaining energy balance. The current study was designed to determine the role of insulin in food intake and its interaction with NPY receptors in 8 experiments using broiler cockerels (4 treatment groups per experiment, except for experiment 8). Chicks received control solution or 2.5, 5, or 10 ng of insulin in experiment 1 and control solution or 1.25, 2.5, or 5 μg of receptor antagonists B5063, SF22, or SML0891 in experiments 2, 3, and 4 through intracerebroventricular (ICV) injection, respectively. In experiments 5, 6, and 7, chicks received ICV injection of B5063, SF22, SML0891, or co-injection of an antagonist + insulin, control solution, and insulin. In experiment 8, blood glucose was measured. Insulin, B5063, and SML0891 decreased food intake, while SF22 led to an increase in food intake. The hypophagic effect of insulin was also reinforced by injection of B560, but ICV injection of SF22 destroyed this hypophagic effect of insulin and increased food intake (p < 0.05). However, SML0891 had no effect on decreased food intake induced by insulin (p > 0.05). At 30 min postinjection, blood sugar in the control group was higher than that in the insulin group (p < 0.05). Therefore, the NPY1 and NPY2 receptors mediate the hypophagic effect of insulin in broiler cockerels.