Merck
  • Dexmedetomidine ameliorates lipopolysaccharide-induced acute kidney injury in rats by inhibiting inflammation and oxidative stress via the GSK-3β/Nrf2 signaling pathway.
Dexmedetomidine ameliorates lipopolysaccharide-induced acute kidney injury in rats by inhibiting inflammation and oxidative stress via the GSK-3β/Nrf2 signaling pathway.
Journal of cellular physiology (2019-03-29)
Xiujing Feng, Wei Guan, Yuan Zhao, Chaoran Wang, Manyu Song, Yujie Yao, Tianyuan Yang, Honggang Fan
RESUMO
Acute kidney injury (AKI) is a frequent and serious complication of sepsis; however, there are currently no effective therapies. Inflammation and oxidative stress are the major mechanisms implicated in lipopolysaccharide (LPS)-induced AKI. Dexmedetomidine (DEX) has been reported to have remarkable anti-inflammatory and antioxidant effects. Here, we examined the renoprotective effects of DEX and potential underlying mechanisms in rats with LPS-induced AKI. We analyzed renal function and structure; serum inflammatory cytokine; renal oxidant and antioxidant levels; and renal expression of glycogen synthase kinase-3β (GSK-3β)/nuclear factor erythroid 2-related factor 2 (Nrf2) pathway-related proteins in rats 4 hr after administration of LPS. Pretreatment with DEX improved renal function and significantly reduced the levels of inflammatory cytokines and oxidative stress markers. Treatment with DEX and the GSK-3β inhibitor SB216367 promoted phosphorylation of GSK-3β, induced Nrf2 nuclear translocation, and increased transcription of the Nrf2 target genes heme oxygenase-1 and NAD(P)H quinone oxidoreductase-1, primarily in renal tubules. Alpha-2-adrenergic receptor (α2-AR) antagonist atipamezole and imidazoline I 2 receptor (I 2 R) antagonist idazoxan reversed the effects of DEX. These results suggest that the renoprotective effects of DEX are mediated via α2-AR and I 2 R-dependent pathways that reduce inflammation and oxidative stress through GSK-3β/Nrf2 signaling.