- A versatile catalyst system for Suzuki-Miyaura cross-coupling reactions of C(sp(2))-tosylates and mesylates.
A versatile catalyst system for Suzuki-Miyaura cross-coupling reactions of C(sp(2))-tosylates and mesylates.
Organic letters (2009-08-12)
Brijesh Bhayana, Brett P Fors, Stephen L Buchwald
PMID19663467
RESUMO
A catalyst system for the Suzuki-Miyaura cross-coupling reactions of aryl and vinyl tosylates and mesylates has been developed. This catalyst displays excellent functional group tolerance and allows the coupling of heteroarylboronic acids with aryl tosylates and mesylates to be performed in high yields. Moreover, reactions employing alkylboronic acids, as well as heteroaryl, vinyl, and allylic pinacol boronate esters, were conducted with high efficiencies.
MATERIAIS
Número do produto
Marca
Descrição do produto