Merck
Synthesis and evaluation of a miniature organic model of chymotrypsin.
Biochemical and biophysical research communications (1985-06-28)
V T D'Souza, K Hanabusa, T O'Leary, R C Gadwood, M L Bender
RESUMO
An artificial chymotrypsin, with all the features of the real chymotrypsin, namely a binding site (from cyclodextrin) attached to a catalytic site containing an imidazolyl group, a carboxylate group and a hydroxyl group, has been synthesized. This artificial chymotrypsin has a molecular weight of only 1,365 while the real enzyme has a molecular weight of 24,800. However, from preliminary measurements, both the real and artificial enzymes have approximately the same catalytic activity (both rate and binding constants).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diethylaminoethyl–Sephacel®, aqueous ethanol suspension, 40-160 μm (wet), exclusion limit ~1,000,000 Da