Merck
  • A novel technique for the effective concentration of 99mTc from a large alumina column loaded with low specific-activity (n,gamma)-produced 99Mo.
A novel technique for the effective concentration of 99mTc from a large alumina column loaded with low specific-activity (n,gamma)-produced 99Mo.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2008-04-05)
Sankha Chattopadhyay, Malay Kanti Das
RESUMO
Molybdenum-99 of low-to-medium specific activity, 7.4-14.8 GBq/g, obtained by the (98)Mo(n,gamma)(99)Mo reaction in research reactors requires a large alumina column to adsorb the molybdenum since the capacity of alumina to adsorb Mo is limited (20 mg Mo/g of alumina). The large size of alumina column, in turn, requires large eluant volumes to elute the (99m)Tc, and the radioactive concentration (RAC) of the pertechnetate becomes unacceptably low for radiopharmaceutical applications. This study describes a method for obtaining high RAC pertechnetate solutions from a column generator loaded with (99)Mo of low specific activity. Molybdenum-99 of specific activity, 7.4 GBq/g, was absorbed on an alumina column [60 g, 70 mm, (H) x 35 mm (diam.)]. The (99m)TcO(4)(-), eluted with 60 ml normal saline (0.15N NaCl), was applied to a 12 mm (H) x 1 mm (diam.) column containing 15 mg of the strong anion exchanger Dowex 1 x 8. The retained pertechnetate was eluted with 5 ml of 0.5 mM tetrabutylammonium bromide (TBAB) solution in methylene chloride. The organic solvent was subsequently removed under vacuum and the dry residue was reconstituted with the desired volume of normal saline. All the operations were carried out in a closed cycle with sterile connecting tubes and multi-way stop-cocks. The concentrated pertechnetate obtained contained about 90% of the total saline eluate and had a pH of 6-7, a radiochemical purity (RCP) consistently >99%, the (99)Mo breakthrough was around 10(-4)%. Colorimetric spot tests (limit tests) for Al and Mo showed <10 ppm in the final (99m)TcO(4)(-) solution. The RCP of the labelled compounds prepared using the pertechnetate obtained and five different radiopharmaceutical kits were >95%. The procedure may be equally applicable for the effective concentration of (188)ReO(4)(-) from (188)W-(188)Re generators.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
AmberChrom 1X2 chloride form, 50-100 mesh