Merck
Chitosan membranes modified by contact with poly(acrylic acid).
Carbohydrate research (2009-06-27)
M S P De Lima, M S Freire, J L C Fonseca, M R Pereira
RESUMO
In this work chitosan membranes modified by contact with poly(acrylic acid) (PAA) aqueous solution at two different temperatures (25 degrees C and 60 degrees C) were obtained. The pure chitosan (CS) membranes, as well as those treated with PAA (CSPAA_25 and CSPAA_60) were characterized by FTIR-ATR, water sorption capacity, thermal analysis (TG/DTG), and scanning electron microscopy (SEM). In addition, in vitro permeation experiments were carried out using metronidazol and sodium sulfamerazine aqueous solutions at 0.1% and 0.2% as model drugs. FTIR-ATR results showed the presence of absorption bands of NH3+ and COO(-) indicating the formation of a polyelectrolyte complex between chitosan and poly(acrylic acid). The results also indicated that PAA penetrates deeper into the membrane at higher temperature (60 degrees C), forming a thicker complex layer. Polyelectrolyte complex formation as well as the influence of treatment temperature was confirmed by lower hydrophilicity, higher thermal stability, and lower permeability of the treated membranes. The results show that the methodology used is a simple and very efficient way to drastically change some membrane properties, especially their permeability.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Supelco
Sulfamerazine, VETRANAL®, analytical standard