Merck
Xylose metabolism in the pig.
PloS one (2018-10-26)
Nichole F Huntley, John F Patience
RESUMO
It is important to understand if, and to what extent, the pig can utilize xylose as an energy source if xylanase releases free xylose in the small intestine. The experimental objectives were to determine the effects of industry-relevant dietary xylose concentrations and adaptation time on xylose retention efficiency and metabolism, diet digestibility and energy value, nitrogen balance, and hindgut fermentation. Forty-eight pigs were housed in metabolism crates and randomly assigned to one of four treatments with increasing D-xylose levels (n = 12/treatment) in 2 replications of a 22-d experiment with 3 collection periods. The control diet was xylose-free (0%), to which either 2, 4, or 8% D-xylose was added. Adaptation effects were assessed during three fecal and urine collection periods: d 5-7, 12-14, and 19-21. On d 22, pigs from the 0 and 8% treatments were euthanized; cecal and colon digesta were collected. Dietary xylose did not affect the total tract digestibility of dry matter, gross energy, or crude protein (P>0.10). Digesta short chain fatty acids concentrations and molar proportions and cecal pH were not different (P>0.10). This experiment utilized a targeted metabolomics approach to characterize and quantify urine xylose and metabolite excretion. Xylose retention decreased from 60% to 47% to 41% when pigs were fed diets containing 2, 4, or 8% xylose, respectively. In the 4 and 8% treatments, xylose retention was greater in the 2nd and 3rd collection periods compared to the 1st. A comprehensive pathway for xylose metabolism was proposed and D-threitol was confirmed as the major urinary metabolite of xylose. In conclusion, pigs can metabolize xylose, but with considerably lower efficiency than glucose, and may be able to adapt with time to utilize xylose more efficiently.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
D-Xylulose, ≥95% (HPLC), syrup
Preço e disponibilidade não estão disponíveis no momento.