Merck
  • Multiple tumor suppressors regulate a HIF-dependent negative feedback loop via ISGF3 in human clear cell renal cancer.
Multiple tumor suppressors regulate a HIF-dependent negative feedback loop via ISGF3 in human clear cell renal cancer.
eLife (2018-10-26)
Lili Liao, Zongzhi Z Liu, Lauren Langbein, Weijia Cai, Eun-Ah Cho, Jie Na, Xiaohua Niu, Wei Jiang, Zhijiu Zhong, Wesley L Cai, Geetha Jagannathan, Essel Dulaimi, Joseph R Testa, Robert G Uzzo, Yuxin Wang, George R Stark, Jianxin Sun, Stephen Peiper, Yaomin Xu, Qin Yan, Haifeng Yang
RESUMO
Whereas VHL inactivation is a primary event in clear cell renal cell carcinoma (ccRCC), the precise mechanism(s) of how this interacts with the secondary mutations in tumor suppressor genes, including PBRM1, KDM5C/JARID1C, SETD2, and/or BAP1, remains unclear. Gene expression analyses reveal that VHL, PBRM1, or KDM5C share a common regulation of interferon response expression signature. Loss of HIF2α, PBRM1, or KDM5C in VHL-/-cells reduces the expression of interferon stimulated gene factor 3 (ISGF3), a transcription factor that regulates the interferon signature. Moreover, loss of SETD2 or BAP1 also reduces the ISGF3 level. Finally, ISGF3 is strongly tumor-suppressive in a xenograft model as its loss significantly enhances tumor growth. Conversely, reactivation of ISGF3 retards tumor growth by PBRM1-deficient ccRCC cells. Thus after VHL inactivation, HIF induces ISGF3, which is reversed by the loss of secondary tumor suppressors, suggesting that this is a key negative feedback loop in ccRCC.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
ANTI-IRF9 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution