Merck
  • Structure-guided design and optimization of small molecules targeting the protein-protein interaction between the von Hippel-Lindau (VHL) E3 ubiquitin ligase and the hypoxia inducible factor (HIF) alpha subunit with in vitro nanomolar affinities.
Structure-guided design and optimization of small molecules targeting the protein-protein interaction between the von Hippel-Lindau (VHL) E3 ubiquitin ligase and the hypoxia inducible factor (HIF) alpha subunit with in vitro nanomolar affinities.
Journal of medicinal chemistry (2014-08-29)
Carles Galdeano, Morgan S Gadd, Pedro Soares, Salvatore Scaffidi, Inge Van Molle, Ipek Birced, Sarah Hewitt, David M Dias, Alessio Ciulli
RESUMO
E3 ubiquitin ligases are attractive targets in the ubiquitin-proteasome system, however, the development of small-molecule ligands has been rewarded with limited success. The von Hippel-Lindau protein (pVHL) is the substrate recognition subunit of the VHL E3 ligase that targets HIF-1α for degradation. We recently reported inhibitors of the pVHL:HIF-1α interaction, however they exhibited moderate potency. Herein, we report the design and optimization, guided by X-ray crystal structures, of a ligand series with nanomolar binding affinities.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(S,R,S)-AHPC hydrochloride, ≥97%