Merck
  • In Vitro Activities of Ceftazidime-Avibactam, Aztreonam-Avibactam, and a Panel of Older and Contemporary Antimicrobial Agents against Carbapenemase-Producing Gram-Negative Bacilli.
In Vitro Activities of Ceftazidime-Avibactam, Aztreonam-Avibactam, and a Panel of Older and Contemporary Antimicrobial Agents against Carbapenemase-Producing Gram-Negative Bacilli.
Antimicrobial agents and chemotherapy (2015-09-24)
Shawn Vasoo, Scott A Cunningham, Nicolynn C Cole, Peggy C Kohner, Sanjay R Menon, Kevin M Krause, Kelly A Harris, Partha P De, Tse Hsien Koh, Robin Patel
RESUMO
Among 177 carbapenemase-producing Gram-negative bacilli (108 KPC, 32 NDM, 11 IMP, 8 OXA-48, 4 OXA-181, 2 OXA-232, 5 IMI, 4 VIM, and 3 SME producers), aztreonam-avibactam was active against all isolates except two NDM producers with elevated MICs of 8/4 and 16/4 mg/liter; ceftazidime-avibactam was active against all KPC-, IMI-, SME-, and most OXA-48 group-producing isolates (93%) but not metallo-β-lactamase producers. Among older and contemporary antimicrobials, the most active were colistin, tigecycline, and fosfomycin, with overall susceptibilities of 88%, 79%, and 78%, respectively.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aztreonam, ≥98% (HPLC)