22742-U
Fritted Purge Sampler
size 5 mL, right stem
, Tekmar® 14-3544-124
About This Item
Materiais
glass
fabricante/nome comercial
Tekmar® 14-3544-124
tamanho
5 mL
aplicação(ões)
industrial hygiene
compatibilidade
for use with Tekmar® 2000/3000
Características e benefícios
Note: All 1/4 in. O.D. glass ends are ground glass.
Informações legais
Tekmar is a registered trademark of Tekmar Co.
Choose from one of the most recent versions:
Certificados de análise (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atendimento ao cliente