Merck
HomeApplicationsProtein BiologyLysis & Protein Extraction
Lysis & Protein Extraction
A digital illustration of multiple blue colored cells, representing human cells, suspended in a light blue environment.
When purifying proteins for functional or structural studies, or for preparative processing and production, the first step is to disrupt the cells or tissue to gain access to the target proteins. Cell lysis and protein solubilization are key to effective analysis and efficient processing. The choice of extraction method can be enzymatic, chemical, mechanical or a combination.
Something went wrong, please try again.
Featured Categories
Cell lysis for protein extraction.
Whole Cell Lysis Reagents & Enhancers
Protein extraction kits, cell lysis buffers, and reagents for solubilizing proteins from bacteria, yeast, and insect cultures, as well as plant and mammalian cell cultures and tissue samples.
Shop Products
A 3D rendering of intricate purple protein molecules floating against a deep blue background. The molecules vary in size and are scattered throughout, with a complex structure featuring multiple protrusions and cavities. They are highlighted by the lighting, giving them a glossy appearance..
Phosphatase and Protease Inhibitor Cocktails
Protease and phosphatase inhibitors and cocktails to prevent proteolysis and protein dephosphorylation, and to preserve the active state of proteins during cell lysis, protein extraction, and sample preparation.
Shop Products
Subcellular fractionation techniques
Subcellular Fractionation, Enrichment, and Depletion Reagents
Uncover proteins with precision: Explore kits for subcellular fractionation, protein enrichment, & extraction across diverse sample types in proteomics research.
Shop Products
Extreme close-up of detergent bubbles in different sizes
Detergents - Anionic, Cationic, Zwitterionic, Anti-foaming
Discover versatile biological detergents, surfactants for lysis, electrophoresis, WB, transfection, research applications. REACH-compliant options available.
Shop Products
Numerous methods are available for disrupting cells and preparing their contents for analysis. In general, gentle methods are employed when the sample consists of easily lysed cultured cells or blood cells, whereas more vigorous methods are employed for the disruption of more robust bacterial or plant cells, or mammalian cells embedded in connective tissue.
  • Detergent-based lysis: Detergent-based lysis: Detergent lysis is a gentle method that can be used for mammalian cells, bacterial cells, yeasts, and plants. Cell suspensions are gently centrifuged and resuspended in lysis solution containing detergents which act to disrupt the cell membrane. The membranes are solubilized, lysing cells and liberating their contents. Detergents may need to be removed downstream if they interfere with analysis or production.
  • Freeze-thaw lysis: This method is applicable to suspensions of mammalian or bacterial cells. The cell suspension is rapidly frozen using liquid nitrogen. The sample is then thawed and resuspended by pipetting or gentle vortexing in lysis buffer, repeating the process several times. Between cycles the sample is centrifuged, and the supernatant containing soluble protein is retained.
  • Osmotic shock: This is a very gentle method that may be sufficient for the lysis of suspended mammalian or bacterial cells without the use of a detergent. The method, often combined with mechanical disruption, relies on changing from high to low osmotic medium, and is well-suited to applications in which the lysate is to be subsequently fractionated into subcellular components.
  • Ultrasonication: This method of protein extraction is most frequently applied to cell suspensions. Cells are disrupted by high-frequency sound waves via a probe inserted in the sample. The sound waves generate a region of low pressure, causing disruption of the cell membranes.
  • Mechanical methods: Proteins may be extracted from cells and tissues using various crude but effective "crushing and grinding" measures. For example, cell membranes may be disrupted by liquid shear forces using Dounce or Potter-Elvehjem homogenization. Tissues may be homogenized by chopping or mincing in chilled buffer using a Waring blender or Polytron® homogenizer. Tissues or cells may be frozen in liquid nitrogen and ground to a fine powder using a mortar and pestle with alumina or sand. Rapid agitation of cells with fine glass beads disrupts cell walls, effective for most Gram positive and Gram negative bacteria.
  • Enzymatic digestion: Enzymatic digestion: Enzymatic methods are frequently used when extracting proteins from bacteria, yeast, or eukaryotic cells embedded in fibrous tissues where cell membranes are surrounded by a robust protective structure. Cell lysis enzymes or cocktails such as Lysozyme, Mutanolysin, MetaPolyzyme, Lysonase, and Pronase can be used in combination with tissue digestion enzymes (i.e. Collagenase, Chondroitinase, Hyaloronidase) to dissolve or disrupt cell walls, coats, capsules, capsids, or other structures not easily sheared by mechanical methods alone. Enzymatic digestion can be followed by homogenization, sonication, or vigorous vortexing in a lysis buffer.
Additionally, because endogenous proteases and phosphatases may be liberated upon cell disruption and degrade the target molecule, the sample should be protected during cell disruption and subsequent purification using protease and phosphatase inhibitors to avoid uncontrolled loss of target.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents