Merck
NIOSH 9000
ASBESTOS, CHRYSOTILE by XRD
This method uses x-ray powder diffraction to perform analysis of percent chrysotile asbestos in bulk samples. Note the precaution that since asbestos is a human carcinogen, it should be handled in a hood. NIOSH methods 7402 (Asbestos by Transmission Electron Microscopy) and 9002 (Asbestos by Polarized Light Microscopy) are also useful for positive identification of asbestos.
Analysis Type (Application Method): X-Ray powder diffraction
Link to NIOSH 9000 Method
Compatible Filters:
25 mm 0.45 μm silver membrane 37 mm 1.0 μm polycarbonate filter (not available) Forceps to handle filter membrane.