Merck
HomeLarge Molecule HPLCHPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution
HPLC Analysis of Immunoglobulin G (IgG) on Zenix® SEC-150 versus Zenix® SEC-300, Effect of Pore Size on Resolution
Graphh Zenix SEC-150 & SEC-300
Conditions
Column   Zenix SEC-150, 30 cm x 4.6 mm I.D., 3 µm, 150 Angstrom particles (Z777016)
Column   Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 µm, 300 Angstrom particles (Z777028)
mobile phase   150 mM sodium phosphate, pH 7.0
Flow rate   1 mL/min
Sample   1 mg/mL, IgG
Injection   10 µL
Detector   UV, 214 nm
Description
Categories Analytical Chromatography, Antibiotics, Antimicrobials
Featured Industry Life Science and Biopharma
Legal Information Zenix is a registered trademark of Sepax Technologies
Suitability application for HPLC
MATERIALS
Sorry, an unexpected error has occurred
Network error: Failed to fetch